2-Amino-5,6-dimethoxypyridine 98%

Supplier: Apollo Scientific

Synonyms: 5,6-Dimethoxypyridin-2-amine

OR909013-1G OR909013-5G
APOSOR909013-1GEA 139 EUR
APOSOR909013-1G APOSOR909013-5G
2-Amino-5,6-dimethoxypyridine 98%
2-Amino-5,6-dimethoxypyridine
Formula: C₇H₁₀N₂O₂
MW: 154,167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09910033
CAS Number: 127980-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR