1-(4-Bromophenyl)cyclopentanecarboxylic acid 97%

Supplier: Apollo Scientific

Synonyms: 1-(4-Bromophenyl)cyclopentane-1-carboxylic acid

OR909248-5G
APOSOR909248-5GEA 242 EUR
APOSOR909248-5G
1-(4-Bromophenyl)cyclopentanecarboxylic acid 97%
1-(4-Bromophenyl)cyclopentanecarboxylic acid
Formula: C₁₂H₁₃BrO₂
MW: 269,134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00455267
CAS Number: 143328-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR