1-(2-Chlorophenyl)cyclopropanecarboxylic acid 97%
Supplier: Apollo Scientific
OR909253-5G
APOSOR909253-5GEA
144
EUR
APOSOR909253-5G
1-(2-Chlorophenyl)cyclopropanecarboxylic acid 97%
1-(2-Chlorophenyl)cyclopropanecarboxylic acid
Formula:
C₁₀H₉ClO₂ MW: 196,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07374435 CAS Number: 122143-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...