N-Cyclopropyl-1H-pyrrole-2-carboxamide 98%
Supplier: Apollo Scientific
OR909323-1G
OR909323-25G
OR909323-5G
APOSOR909323-1GEA
125
EUR
APOSOR909323-1G
APOSOR909323-25G
APOSOR909323-5G
N-Cyclopropyl-1H-pyrrole-2-carboxamide 98%
N-Cyclopropyl-1H-pyrrole-2-carboxamide
Formula:
C₈H₁₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD15143557 CAS Number: 1215206-52-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...