5-Chloro-6-methylbenzoimidazol-2-one 95%

Supplier: Apollo Scientific

OR909349-1G OR909349-5G
APOSOR909349-1GEA 328 EUR
APOSOR909349-1G APOSOR909349-5G
5-Chloro-6-methylbenzoimidazol-2-one 95%
5-Chloro-6-methylbenzoimidazol-2-one
Formula: C₈H₇ClN₂O
MW: 182,607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15143569
CAS Number: 683240-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR