7-Chloro-benzofuran 97%

Supplier: Apollo Scientific
Warning

Synonyms: 7-Chlorobenzofuran

OR909406-1G OR909406-5G
APOSOR909406-1GEA 55.1 EUR
APOSOR909406-1G APOSOR909406-5G
7-Chloro-benzofuran 97%
7-Chloro-benzofuran
Formula: C₈H₅ClO
MW: 152,578 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18207570
CAS Number: 24410-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR