Ethyl pyrrole-1-carboxylate 98%

Supplier: Apollo Scientific

OR909473-1G OR909473-5G
APOSOR909473-1GEA 132 EUR
APOSOR909473-1G APOSOR909473-5G
Ethyl pyrrole-1-carboxylate 98%
Ethyl pyrrole-1-carboxylate
Formula: C₇H₉NO₂
MW: 139,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12923844
CAS Number: 4277-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR