1-Propyl-3H-1,3-benzodiazol-2-one 97%

Supplier: Apollo Scientific

OR909488-10G OR909488-1G OR909488-5G
APOSOR909488-10GEA 754 EUR
APOSOR909488-10G APOSOR909488-1G APOSOR909488-5G
1-Propyl-3H-1,3-benzodiazol-2-one 97%
1-Propyl-3H-1,3-benzodiazol-2-one
Formula: C₁₀H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD02707887
CAS Number: 77557-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR