2-Pentanoyl-3-picoline 96%
Supplier: Apollo Scientific
OR909610-1G
OR909610-5G
APOSOR909610-1GEA
219
EUR
APOSOR909610-1G
APOSOR909610-5G
2-Pentanoyl-3-picoline 96%
2-Pentanoyl-3-picoline
Formula:
C₁₁H₁₅NO MW: 177.243 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16820170 CAS Number: 1249752-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...