4-Bromo-1-(methoxymethyl)pyrazole 98%

Supplier: Apollo Scientific

Synonyms: 4-Bromo-1-(methoxymethyl)-1H-pyrazole

OR909723-1G OR909723-5G
APOSOR909723-1GEA 219 EUR
APOSOR909723-1G APOSOR909723-5G
4-Bromo-1-(methoxymethyl)pyrazole 98%
4-Bromo-1-(methoxymethyl)pyrazole
Formula: C₅H₇BrN₂O
MW: 191,026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18324047
CAS Number: 1071200-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR