2-Chloro-N-ethylpyridin-5-amine 98%
Supplier: Apollo Scientific
OR909748-1G
OR909748-5G
APOSOR909748-1GEA
290
EUR
APOSOR909748-1G
APOSOR909748-5G
2-Chloro-N-ethylpyridin-5-amine 98%
2-Chloro-N-ethylpyridin-5-amine
Formula:
C₇H₉ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD24255474 CAS Number: 120739-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...