7-methylimidazo[1,2-a]pyridine-3-carbaldehyde 95%

Supplier: Apollo Scientific

OR909942-1G OR909942-250MG
APOSOR909942-1GEA 735 EUR
APOSOR909942-1G APOSOR909942-250MG
7-methylimidazo[1,2-a]pyridine-3-carbaldehyde 95%
7-methylimidazo[1,2-a]pyridine-3-carbaldehyde
Formula: C₉H₈N₂O
MW: 160,173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09994349
CAS Number: 30384-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR