Methyl 1-methylbenzimidazole-6-carboxylate 97%

Supplier: Apollo Scientific

OR910155-1G OR910155-5G
APOSOR910155-1GEA 112 EUR
APOSOR910155-1G APOSOR910155-5G
Methyl 1-methylbenzimidazole-6-carboxylate 97%
Methyl 1-methylbenzimidazole-6-carboxylate
Formula: C₁₀H₁₀N₂O₂
MW: 190,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11973677
CAS Number: 53484-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR