3-(4-Chlorophenyl)-5-methyl-1,2,4-oxadiazole 96%
Supplier: Apollo Scientific
OR910206-1G
OR910206-25G
OR910206-5G
APOSOR910206-1GEA
125
EUR
APOSOR910206-1G
APOSOR910206-25G
APOSOR910206-5G
3-(4-Chlorophenyl)-5-methyl-1,2,4-oxadiazole 96%
3-(4-Chlorophenyl)-5-methyl-1,2,4-oxadiazole
Formula:
C₉H₇ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD00086978 CAS Number: 21614-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...