6-Bromo-2-chloroimidazo[1,2-a]pyridine 95+%
Supplier: Apollo Scientific
|
Warning
|
OR910232-1G
OR910232-250MG
APOSOR910232-1GEA
144
EUR
APOSOR910232-1G
APOSOR910232-250MG
6-Bromo-2-chloroimidazo[1,2-a]pyridine 95+%
6-Bromo-2-chloroimidazo[1,2-a]pyridine
Formula:
C₇H₄BrClN₂ MW: 231,477 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11976188 CAS Number: 851916-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...