Methyl 2-methylpyrazole-3-carboxylate 97%

Supplier: Apollo Scientific

Synonyms: Methyl 1-methyl-1H-pyrazole-5-carboxylate

OR910254-10G OR910254-1G OR910254-5G
APOSOR910254-10GEA 164 EUR
APOSOR910254-10G APOSOR910254-1G APOSOR910254-5G
Methyl 2-methylpyrazole-3-carboxylate 97%
Methyl 2-methylpyrazole-3-carboxylate
Formula: C₆H₈N₂O₂
MW: 140,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04967357
CAS Number: 17827-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR