N,3-Dimethylthiophene-2-carboxamide 98%
Supplier: Apollo Scientific
OR910356-100MG
OR910356-1G
OR910356-250MG
APOSOR910356-100MGEA
161
EUR
APOSOR910356-100MG
APOSOR910356-1G
APOSOR910356-250MG
N,3-Dimethylthiophene-2-carboxamide 98%
N,3-Dimethylthiophene-2-carboxamide
Formula:
C₇H₉NOS MW: 155,217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11726152 CAS Number: 56776-68-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...