Methyl 4-cyanothiophene-2-carboxylate 97%

Supplier: Apollo Scientific

OR910411-1G OR910411-250MG
APOSOR910411-1GEA 677 EUR
APOSOR910411-1G APOSOR910411-250MG
Methyl 4-cyanothiophene-2-carboxylate 97%
Methyl 4-cyanothiophene-2-carboxylate
Formula: C₇H₅NO₂S
MW: 167,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664843
CAS Number: 67808-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR