5-Methoxy-1-methylindazole 98%

Supplier: Apollo Scientific

OR910414-1G OR910414-250MG
APOSOR910414-1GEA 806 EUR
APOSOR910414-1G APOSOR910414-250MG
5-Methoxy-1-methylindazole 98%
5-Methoxy-1-methylindazole
Formula: C₉H₁₀N₂O
MW: 162,189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22380221
CAS Number: 756839-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR