7-Bromo-2,3-dihydro-1H-indole 95%
Supplier: Apollo Scientific
Synonyms:
7-Bromoindoline
OR910527-1G
OR910527-250MG
APOSOR910527-1GEA
203
EUR
APOSOR910527-1G
APOSOR910527-250MG
7-Bromo-2,3-dihydro-1H-indole 95%
7-Bromo-2,3-dihydro-1H-indole
Formula:
C₈H₈BrN MW: 198,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09731059 CAS Number: 62813-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...