1,2-Dimethyl-1,3-benzodiazole-5-carbonitrile 98%

Supplier: Apollo Scientific

OR910553-1G OR910553-25G OR910553-5G
APOSOR910553-1GEA 145 EUR
APOSOR910553-1G APOSOR910553-25G APOSOR910553-5G
1,2-Dimethyl-1,3-benzodiazole-5-carbonitrile 98%
1,2-Dimethyl-1,3-benzodiazole-5-carbonitrile
Formula: C₁₀H₉N₃
MW: 171,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027483
CAS Number: 80073-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR