1-Ethyl-1,2,3-benzotriazole-5-carbonitrile 98%

Supplier: Apollo Scientific

OR910556-1G OR910556-25G OR910556-5G
APOSOR910556-1GEA 145 EUR
APOSOR910556-1G APOSOR910556-25G APOSOR910556-5G
1-Ethyl-1,2,3-benzotriazole-5-carbonitrile 98%
1-Ethyl-1,2,3-benzotriazole-5-carbonitrile
Formula: C₉H₈N₄
MW: 172,187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13881240
CAS Number: 1119505-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR