N-(4-Methylphenyl)pyrimidin-2-amine 98%

Supplier: Apollo Scientific

OR911119-10G OR911119-1G OR911119-5G
APOSOR911119-10GEA 754 EUR
APOSOR911119-10G APOSOR911119-1G APOSOR911119-5G
N-(4-Methylphenyl)pyrimidin-2-amine 98%
N-(4-Methylphenyl)pyrimidin-2-amine
Formula: C₁₁H₁₁N₃
MW: 185,225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13412781
CAS Number: 198711-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR