2-(Pyrazol-1-ylmethyl)benzonitrile 96%
Supplier: Apollo Scientific
OR911139-10G
OR911139-1G
OR911139-5G
APOSOR911139-10GEA
754
EUR
APOSOR911139-10G
APOSOR911139-1G
APOSOR911139-5G
2-(Pyrazol-1-ylmethyl)benzonitrile 96%
2-(Pyrazol-1-ylmethyl)benzonitrile
Formula:
C₁₁H₉N₃ MW: 183,209 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09863321 CAS Number: 875740-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...