2-Chloro-6-(1-piperazinyl)pyrazine 98%
Supplier: Apollo Scientific
OR911807-1G
OR911807-5G
APOSOR911807-1GEA
190
EUR
APOSOR911807-1G
APOSOR911807-5G
2-Chloro-6-(1-piperazinyl)pyrazine 98%
2-Chloro-6-(1-piperazinyl)pyrazine
Formula:
C₈H₁₁ClN₄ MW: 198,653 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04975444 CAS Number: 64022-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...