2-(4-Chlorophenoxy)propanoic acid 97%

Supplier: Apollo Scientific

OR912090-5G
APOSOR912090-5GEA 48 EUR
APOSOR912090-5G
2-(4-Chlorophenoxy)propanoic acid 97%
2-(4-Chlorophenoxy)propanoic acid
Formula: C₉H₉ClO₃
MW: 200,619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002647
CAS Number: 3307-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR