N,N-Dimethyl-3-nitrobenzamide 98%

Supplier: Apollo Scientific

OR912541-10G OR912541-1G OR912541-5G
APOSOR912541-10GEA 502 EUR
APOSOR912541-10G APOSOR912541-1G APOSOR912541-5G
N,N-Dimethyl-3-nitrobenzamide 98%
N,N-Dimethyl-3-nitrobenzamide
Formula: C₉H₁₀N₂O₃
MW: 194,187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00466214
CAS Number: 7291-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR