3-Cyano-2-methylindole 95%

Supplier: Apollo Scientific

Synonyms: 2-Methyl-1H-indole-3-carbonitrile

OR912676-1G OR912676-250MG
APOSOR912676-1GEA 62.5 EUR
APOSOR912676-1G APOSOR912676-250MG
3-Cyano-2-methylindole 95%
3-Cyano-2-methylindole
Formula: C₁₀H₈N₂
MW: 156,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01475745
CAS Number: 51072-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR