3-Chloro-7-nitroindole 95%
Supplier: Apollo Scientific
OR912798-1G
OR912798-250MG
APOSOR912798-1GEA
516
EUR
APOSOR912798-1G
APOSOR912798-250MG
3-Chloro-7-nitroindole 95%
3-Chloro-7-nitroindole
Formula:
C₈H₅ClN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD02684149 CAS Number: 165669-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...