2,3-Dihydro-1,1-dioxo-1,2-benzisothiazole 95%
Supplier: Apollo Scientific
OR912878-1G
OR912878-250MG
APOSOR912878-1GEA
251
EUR
APOSOR912878-1G
APOSOR912878-250MG
2,3-Dihydro-1,1-dioxo-1,2-benzisothiazole 95%
2,3-Dihydro-1,1-dioxo-1,2-benzisothiazole
Formula:
C₇H₇NO₂S Storage Temperature: Ambient |
MDL Number:
MFCD03426201 CAS Number: 936-16-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...