3-(1H-1,2,4-Triazol-1-yl)benzylamine 97%

Supplier: Apollo Scientific
Danger

Synonyms: 1-[3-(Aminomethyl)phenyl]-1H-1,2,4-triazole, [3-(1H-1,2,4-Triazol-1-yl)phenyl]methylamine

OR9129-1G OR9129-250MG
APOSOR9129-1GEA 1250 EUR
APOSOR9129-1G APOSOR9129-250MG
3-(1H-1,2,4-Triazol-1-yl)benzylamine 97%
3-(1H-1,2,4-Triazol-1-yl)benzylamine
Formula: C₉H₁₀N₄
MW: 174,203 g/mol
Boiling Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD08271898
CAS Number: 741717-66-8
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR