6-Hydroxy-1,3-dihydro-indol-2-one 98%

Supplier: Apollo Scientific

OR913072-1G OR913072-250MG
APOSOR913072-1GEA 452 EUR
APOSOR913072-1G APOSOR913072-250MG
6-Hydroxy-1,3-dihydro-indol-2-one 98%
6-Hydroxy-1,3-dihydro-indol-2-one
Formula: C₈H₇NO₂
MW: 149,147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663501
CAS Number: 6855-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR