4-Chloro-6-ethyl-2-methylpyrimidine 95%

Supplier: Apollo Scientific

OR913279-1G OR913279-250MG
APOSOR913279-1GEA 633 EUR
APOSOR913279-1G APOSOR913279-250MG
4-Chloro-6-ethyl-2-methylpyrimidine 95%
4-Chloro-6-ethyl-2-methylpyrimidine
Formula: C₇H₉ClN₂
MW: 156,613 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782027
CAS Number: 89966-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR