2-Ethylamino-3-nitropyridine 98%

Supplier: Apollo Scientific

OR913365-1G OR913365-5G
APOSOR913365-1GEA 168 EUR
APOSOR913365-1G APOSOR913365-5G
2-Ethylamino-3-nitropyridine 98%
2-Ethylamino-3-nitropyridine
Formula: C₇H₉N₃O₂
MW: 167.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08692122
CAS Number: 26820-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR