4-Chlorothieno[2,3-b]pyridine-5-carbonitrile 95%

Supplier: Apollo Scientific

OR913377-1G OR913377-250MG
APOSOR913377-1GEA 864 EUR
APOSOR913377-1G APOSOR913377-250MG
4-Chlorothieno[2,3-b]pyridine-5-carbonitrile 95%
4-Chlorothieno[2,3-b]pyridine-5-carbonitrile
Formula: C₈H₃ClN₂S
Storage Temperature: Ambient
MDL Number: MFCD08706210
CAS Number: 63873-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR