2-Chloro-5-ethoxypyridine 95%

Supplier: Apollo Scientific

OR913408-1G OR913408-250MG
APOSOR913408-1GEA 155 EUR
APOSOR913408-1G APOSOR913408-250MG
2-Chloro-5-ethoxypyridine 95%
2-Chloro-5-ethoxypyridine
Formula: C₇H₈ClNO
MW: 157,598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835252
CAS Number: 856851-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR