(2-Formylphenoxy)acetonitrile 97%

Supplier: Apollo Scientific

Synonyms: 2-(2-Formylphenoxy)acetonitrile

OR913452-1G OR913452-5G
APOSOR913452-1GEA 60.9 EUR
APOSOR913452-1G APOSOR913452-5G
(2-Formylphenoxy)acetonitrile 97%
(2-Formylphenoxy)acetonitrile
Formula: C₉H₇NO₂
MW: 161,157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01157218
CAS Number: 125418-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR