(R)-2-Acetylamino-3-cyclopropylpropionic acid 95%

Supplier: Apollo Scientific

OR913649-1G OR913649-250MG
APOSOR913649-1GEA 698 EUR
APOSOR913649-1G APOSOR913649-250MG
(R)-2-Acetylamino-3-cyclopropylpropionic acid 95%
(R)-2-Acetylamino-3-cyclopropylpropionic acid
Formula: C₈H₁₃NO₃
MW: 171,194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878740
CAS Number: 121786-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR