Methyl (3R)-piperidine-3-carboxylate 96%

Supplier: Apollo Scientific
Warning

Synonyms: (R)-Methyl piperidine-3-carboxylate

OR913753-1G OR913753-5G
APOSOR913753-1GEA 237 EUR
APOSOR913753-1G APOSOR913753-5G
Methyl (3R)-piperidine-3-carboxylate 96%
Methyl (3R)-piperidine-3-carboxylate
Formula: C₇H₁₃NO₂
MW: 143,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09749870
CAS Number: 164323-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR