(S)-(1-Benzyl-pyrrolidin-3-yl)-methanol 95%

Supplier: Apollo Scientific

Synonyms: (S)-(1-Benzylpyrrolidin-3-yl)methanol

OR913789-1G OR913789-250MG
APOSOR913789-1GEA 116 EUR
APOSOR913789-1G APOSOR913789-250MG
(S)-(1-Benzyl-pyrrolidin-3-yl)-methanol 95%
(S)-(1-Benzyl-pyrrolidin-3-yl)-methanol
Formula: C₁₂H₁₇NO
MW: 191,269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09608006
CAS Number: 78914-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR