(S)-3-Aminomethyl-1-benzylpyrrolidine 95%

Supplier: Apollo Scientific

OR913943-1G OR913943-250MG
APOSOR913943-1GEA 575 EUR
APOSOR913943-1G APOSOR913943-250MG
(S)-3-Aminomethyl-1-benzylpyrrolidine 95%
(S)-3-Aminomethyl-1-benzylpyrrolidine
Formula: C₁₂H₁₈N₂
MW: 190,285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369980
CAS Number: 229323-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR