1-(4-Chlorophenyl)-3-hydroxy-1H-pyrazole 95%

Supplier: Apollo Scientific

Synonyms: 1-(4-Chlorophenyl)-1H-pyrazol-3-ol

OR914118-100G OR914118-25G
APOSOR914118-100GEA 118 EUR
APOSOR914118-100G APOSOR914118-25G
1-(4-Chlorophenyl)-3-hydroxy-1H-pyrazole 95%
1-(4-Chlorophenyl)-3-hydroxy-1H-pyrazole
Formula: C₉H₇ClN₂O
MW: 194,618 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08459260
CAS Number: 76205-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR