1-(4-Methylphenyl)-1H-pyrrole 95%

Supplier: Apollo Scientific

Synonyms: 1-(p-Tolyl)-1H-pyrrole

OR914123-1G OR914123-5G
APOSOR914123-1GEA 219 EUR
APOSOR914123-1G APOSOR914123-5G
1-(4-Methylphenyl)-1H-pyrrole 95%
1-(4-Methylphenyl)-1H-pyrrole
Formula: C₁₁H₁₁N
MW: 157,212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00799565
CAS Number: 827-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR