5-Methoxy-2,4-dihydro-1,4-benzoxazin-3-one 98%
Supplier: Apollo Scientific
OR914310-100MG
OR914310-1G
OR914310-250MG
APOSOR914310-100MGEA
203
EUR
APOSOR914310-100MG
APOSOR914310-1G
APOSOR914310-250MG
5-Methoxy-2,4-dihydro-1,4-benzoxazin-3-one 98%
5-Methoxy-2,4-dihydro-1,4-benzoxazin-3-one
Formula:
C₉H₉NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD11878410 CAS Number: 1058704-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...