3,5-Bis(methylthio)pyridine 97%

Supplier: Apollo Scientific

OR914473-1G OR914473-250MG
APOSOR914473-1GEA 364 EUR
APOSOR914473-1G APOSOR914473-250MG
3,5-Bis(methylthio)pyridine 97%
3,5-Bis(methylthio)pyridine
Formula: C₇H₉NS₂
MW: 171,283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015800
CAS Number: 70999-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR