1-(Triisopropylsilyl)pyrrole-3-carbaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-tri(Propan-2-yl)silylpyrrole-3-carbaldehyde
OR915073-1G
OR915073-5G
APOSOR915073-1GEA
241
EUR
APOSOR915073-1G
APOSOR915073-5G
1-(Triisopropylsilyl)pyrrole-3-carbaldehyde
1-(Triisopropylsilyl)pyrrole-3-carbaldehyde
Formula:
C₁₄H₂₅NOSi MW: 251.44 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28515390 CAS Number: 90971-76-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...