3-Bromo-5-hydroxybenzaldehyde
Supplier: Apollo Scientific
|
Warning
|
OR916487-5G
APOSOR916487-5GEA
220
EUR
APOSOR916487-5G
3-Bromo-5-hydroxybenzaldehyde
3-Bromo-5-hydroxybenzaldehyde
Formula:
C₇H₅BrO₂ MW: 201.017 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06797981 CAS Number: 199177-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...