4-Methylresorcinol 95%

Supplier: Apollo Scientific

Synonyms: 4-Methylbenzene-1,3-diol, 2,4-Dihydroxytoluene

OR916542-1G OR916542-5G
APOSOR916542-1GEA 77.1 EUR
APOSOR916542-1G APOSOR916542-5G
4-Methylresorcinol 95%
4-Methylresorcinol
Formula: C₇H₈O₂
MW: 124,137 g/mol
Boiling Pt: 264 °C
Melting Pt: 104…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016452
CAS Number: 496-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR