N,N-Dipropylpyridin-4-amine 95%
Supplier: Apollo Scientific
OR916862-1G
OR916862-250MG
APOSOR916862-1GEA
844
EUR
APOSOR916862-1G
APOSOR916862-250MG
N,N-Dipropylpyridin-4-amine 95%
N,N-Dipropylpyridin-4-amine
Formula:
C₁₁H₁₈N₂ MW: 178,274 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12913578 CAS Number: 69008-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...