[4-(Pyridin-2-yl)phenyl]methanol 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-[4-(Hydroxymethyl)phenyl]pyridine, 4-(Pyridin-2-yl)benzyl alcohol

OR9172-1G
APOSOR9172-1GEA 318 EUR
APOSOR9172-1G
[4-(Pyridin-2-yl)phenyl]methanol 97%
[4-(Pyridin-2-yl)phenyl]methanol
Formula: C₁₂H₁₁NO
MW: 185,222 g/mol
Melting Pt: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD04116292
CAS Number: 98061-39-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR